1,3-Diphenylacetone ≥99%

Synonyms: 1,3-Diphenylpropan-2-one, 1,3-Diphenyl-2-propanone

D204609-25G D204609-100G
220120-042EA 57.08 USD
220120-042 220120-041
1,3-Diphenylacetone ≥99%
1,3-Diphenylacetone
Formula: (C₆H₅)CH₂C(O)CH₂(C₆H₅)
MW: 210.28 g/mol
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: 30 - 34 °C
Density: 1.195 g/cm³ (0 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00004795
CAS Number: 102-04-5
EINECS: 203-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR