1,1-Diphenylacetone ≥98%

Synonyms: asym. Diphenylacetone, 1,1-Diphenyl-2-propanone

D204404-100G D204404-25G
220120-038EA 274.62 USD
220120-038 220120-039
1,1-Diphenylacetone ≥98%
1,1-Diphenylacetone
Formula: (C₆H₅)₂CHC(O)CH₃
MW: 210.28 g/mol
Boiling Pt: 111 - 126 °C (0.1 -rr)
Melting Pt: 59 - 62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008759
CAS Number: 781-35-1
EINECS: 212-307-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7eb18cea6b136cdea03e91f1525c902f74d1578&vid=aabdc13746192c136e72ece83402aebd3b86fb9d&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR