Diphenylacetic acid ≥99%

Warning

Synonyms: 2,2-Diphenylacetic acid

D204307-5G D204307-100G
220120-469EA 22.62 USD
220120-469 220121-496
Diphenylacetic acid ≥99%
Diphenylacetic acid
Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 142 - 143 °C
Density: 1.258 g/cm³ (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004251
CAS Number: 117-34-0
EINECS: 204-185-0
Merck Index: 13,03348

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR