2,6-Dinitrotoluene ≥98%

Danger

D200603-25G D200603-100G
220121-485EA 100.37 USD
220121-485 220121-484
2,6-Dinitrotoluene ≥98%
2,6-Dinitrotoluene
Formula: C₇H₆N₂O₄
MW: 182.14 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 65.2 °C
Density: 1.538 g/cm³ (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007158
CAS Number: 606-20-2
EINECS: 210-106-0
UN: 3454
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR