1,3-Dinitrobenzene ≥97%

Danger

D194255-25G D194255-100G
220120-965EA 43.83 USD
220120-965 220120-964
1,3-Dinitrobenzene ≥97%
1,3-Dinitrobenzene
Formula: C₆H₄N₂O₄
MW: 168.11 g/mol
Boiling Pt: 276 °C (1013 hPa)
Melting Pt: 90 °C
Density: 1.55 g/cm³ (20 °C)
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00007222
CAS Number: 99-65-0
EINECS: 202-776-8
UN: 3443
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR