2,4-Dinitrobenzaldehyde ≥97%

Warning

D193607-1G D193607-25G D193607-5G
220120-959EA 28.32 USD
220120-959 220120-960 220120-961
2,4-Dinitrobenzaldehyde ≥97%
2,4-Dinitrobenzaldehyde
Formula: C₇H₄N₂O₅
MW: 196.12 g/mol
Boiling Pt: 378 °C (1013 hPa)
Melting Pt: 72 °C
MDL Number: MFCD00013376
CAS Number: 528-75-6
EINECS: 208-440-7
Merck Index: 13,03300

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR