2,4-Diamino-6-hydroxypyrimidine

Synonyms: 6-Aminoisocytosin, 2,4-Diamino-6-pyrimidinol, 2,4-Diaminopyrimidin-6-ol

D19206-25G D19206-100G
220120-950EA 220.8 USD
220120-950 220121-470
2,4-Diamino-6-hydroxypyrimidine
2,4-Diamino-6-hydroxypyrimidine
Formula: C₄H₆N₄O
MW: 126.12 g/mol
Boiling Pt: 373 °C (1013 hPa)
Melting Pt: 285…286 °C
MDL Number: MFCD00006098
CAS Number: 56-06-4
EINECS: 200-254-4
Merck Index: 13,03001

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR