N,N-Dimethyl-p-toluidine ≥99%

Danger

Synonyms: N,N,4-Trimethylaniline, N,N-Dimethyl-4-methylaniline, 1-(Dimethylamino)-4-methylbenzol, 4,N,N-Trimethylaniline, 4-(Dimethylamino)toluene, 4-Dimethylaminotoluene

D189006-1L D189006-500ML D189006-100ML
220121-465EA 269.23 USD
220121-465 220121-466 220121-464
N,N-Dimethyl-p-toluidine ≥99%
N,N-Dimethyl-p-toluidine
Formula: C₉H₁₃N
MW: 135.21 g/mol
Boiling Pt: 211 °C (1013 hPa)
Melting Pt: -15 °C
Density: 0.9 g/cm³ (20 °C)
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00008316
CAS Number: 99-97-8
EINECS: 202-805-4
UN: 1708
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR