2,6-Dimethyl-4-pyrone

Synonyms: 2,6-Dimethyl-4H-pyran-4-one, 2,6-Dimethyl-γ-pyrone

D183407-100G D183407-25G
220121-430EA 949.85 USD
220121-430 220121-431
2,6-Dimethyl-4-pyrone
2,6-Dimethyl-4-pyrone
Formula: C₇H₈O₂
MW: 124.14 g/mol
Boiling Pt: 248 °C (1013 hPa)
Melting Pt: 130 - 131 °C
Density: 1.17 g/cm³ (25 °C)
MDL Number: MFCD00006579
CAS Number: 1004-36-0
EINECS: 213-719-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR