4,5-Dimethyl-o-phenylenediamine ≥98%

Synonyms: 4,5-Dimethylbenzene-1,2-diamine, 4,5-Dimethyl-1,2-phenylenediamine, 4,5-Diamino-1,2-dimethylbenzene, 1,2-Diamino-4,5-dimethylbenzene

D176605-5G D176605-25G
220121-310EA 119.54 USD
220121-310 220121-309
4,5-Dimethyl-o-phenylenediamine ≥98%
4,5-Dimethyl-o-phenylenediamine
Formula: C₈H₁₂N₂
MW: 136.2 g/mol
MDL Number: MFCD00007729
CAS Number: 3171-45-7
EINECS: 221-635-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR