2,5-Xylenol ≥99%

Danger

Synonyms: 2,5-Dimethylphenol, 2-Hydroxy-p-xylene, 2-Hydroxy-4-xylene

D174602-100G D174602-5G
220121-293EA 64.62 USD
220121-293 220121-294
2,5-Xylenol ≥99%
2,5-Xylenol
Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 212 °C (1013 hPa)
Melting Pt: 75 °C
Density: 1.169 g/cm³ (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002237
CAS Number: 95-87-4
EINECS: 202-461-5
UN: 2261
ADR: 6.1,II
Merck Index: 12,10215

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR