2,4-Xylenol ≥98%

Danger

Synonyms: 4-Hydroxy-3-xylene, 2,4-Dimethylphenol, 2,4-Xylenol

D174203-25ML D174203-100ML
220121-292EA 33.17 USD
220121-292 220121-291
2,4-Xylenol ≥98%
2,4-Xylenol
Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 210.9 °C (1013 hPa)
Melting Pt: 25 °C
Density: 1.016 g/cm³ (25 °C)
Flash Pt: 80 - 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00002233
CAS Number: 105-67-9
EINECS: 203-321-6
UN: 2261
ADR: 6.1,II
Merck Index: 12,10215

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR