2,4-Dimethylpentane ≥99%

Danger

D173401-5G D173401-25G
220121-285EA 126 USD
220121-285 220121-284
2,4-Dimethylpentane ≥99%
2,4-Dimethylpentane
Formula: C₇H₁₆
MW: 100.2 g/mol
Boiling Pt: 81 °C (1013 hPa)
Melting Pt: –119.2 °C
Density: 0.6734 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008945
CAS Number: 108-08-7
EINECS: 203-548-0
UN: 1206
ADR: 3,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR