1,4-Dimethylnaphthalene

D170305-5G D170305-25G
220121-273EA 188.46 USD
220121-273 220121-272
1,4-Dimethylnaphthalene
1,4-Dimethylnaphthalene
Formula: C₁₂H₁₂
MW: 156.23 g/mol
Boiling Pt: 264 °C (1013 hPa)
Density: 1.016 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004037
CAS Number: 571-58-4
EINECS: 209-335-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR