Dimethyl methylphosphonate

D169102-100G D169102-500G D169102-5G
220121-263EA 59.98 USD
220121-263 220121-264 220121-265
Dimethyl methylphosphonate
Dimethyl methylphosphonate
Formula: C₃H₉O₃P
MW: 124.08 g/mol
Boiling Pt: 181 °C (1013 hPa)
Density: 1.1596 g/cm³ (20 °C)
MDL Number: MFCD00008349
CAS Number: 756-79-6
EINECS: 212-052-3
Merck Index: 13,03425

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR