1,12-Diaminododecane ≥98%

Danger

Synonyms: N12N, Dodecamethylendiamine, 1,12-Dodecanediamine

D16401-25G D16401-5G
220121-238EA 103.28 USD
220121-238 220121-239
1,12-Diaminododecane ≥98%
1,12-Diaminododecane
Formula: C₁₂H₂₈N₂
MW: 200.37 g/mol
Boiling Pt: 275 °C (1013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00008155
CAS Number: 2783-17-7
EINECS: 220-489-6
UN: 3259
ADR: 8,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR