N,N-Dimethylhydrazine ≥98%

Synonyms: 1,1-Dimethylhydrazine

D161608-500G D161608-5G D161608-100G
220121-221EA 351.08 USD
220121-221 220121-222 220121-220
N,N-Dimethylhydrazine ≥98%
N,N-Dimethylhydrazine
Formula: C₂H₈N₂
MW: 60.1 g/mol
CAS Number: 57-14-7
EINECS: 200-316-0
UN: 1163
ADR: 6.1,I

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR