N,N-Dimethylethylendiamine ≥95%

Danger

Synonyms: N,N-Dimethylethylenediamine, asym. Dimethylethylendiamine, unsym-Dimethylethylenediamine, 2-(Dimethylamino)ethylamine, 1-Amino-2-dimethylaminoethane, 2-Dimethylaminoethylamine, 2-Aminoethyldimethylamine

D158003-100G D158003-25G
220121-143EA 145.38 USD
220121-143 220121-144
N,N-Dimethylethylendiamine ≥95%
N,N-Dimethylethylendiamine
Formula: C₄H₁₂N₂
MW: 88.15 g/mol
Boiling Pt: ∼106 °C (1013 hPa)
Melting Pt: –70 °C
Density: 0.804 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00008175
CAS Number: 108-00-9
EINECS: 203-541-2

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR