N,N'-Dimethylethylendiamine ≥99%

Synonyms: N1,N2-Dimethylethane-1,2-diamine, sym. Dimethylethylendiamine, N,N'-Dimethylethylenediamine

D157805-25G D157805-5G D157805-100G
220121-141EA 147.54 USD
220121-141 220121-142 220121-140
N,N'-Dimethylethylendiamine ≥99%
N,N'-Dimethylethylendiamine
Formula: C₄H₁₂N₂
MW: 88.15 g/mol
Boiling Pt: 119 °C (1013 hPa)
Density: 0.82 g/cm³ (20 °C)
MDL Number: MFCD00008290
CAS Number: 110-70-3
EINECS: 203-793-3

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR