3,4-Dimethylbenzoic acid ≥98%

D149403-10G D149403-100G
220120-639EA 52.03 USD
220120-639 220120-638
3,4-Dimethylbenzoic acid ≥98%
3,4-Dimethylbenzoic acid
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 295 °C (1013 hPa)
Melting Pt: 163…164 °C
Storage Temperature: Ambient
MDL Number: MFCD00002524
CAS Number: 619-04-5
EINECS: 210-576-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR