2,6-Dimethylanisole ≥98%

D146404-100G D146404-25G
220120-630EA 216.46 USD
220120-630 220120-631
2,6-Dimethylanisole ≥98%
2,6-Dimethylanisole
Formula: C₉H₁₂O
MW: 136.19 g/mol
Boiling Pt: 182 °C (1013 hPa)
Density: 0.96 g/cm³ (20 °C)
MDL Number: MFCD00008380
CAS Number: 1004-66-6
EINECS: 213-723-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR