2,6-Xylidine ≥99%

Warning

Synonyms: 2-Amino-m-xylene, 2,6-Dimethylaniline

D146005-5G D146005-500G D146005-100G
220120-629EA 49.32 USD
220120-629 220120-628 220120-627
2,6-Xylidine ≥99%
2,6-Xylidine
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 214 °C (1013 hPa)
Melting Pt: 10 - 12 °C
Density: 0.98 g/cm³ (20 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00007747
CAS Number: 87-62-7
EINECS: 201-758-7
UN: 1711
ADR: 6.1,II
Merck Index: 12,10217

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR