2,3-Dimethylaniline ≥99%

Danger

Synonyms: 2,3-Xylidine, 3-Amino-o-xylene

D145807-100G D145807-500G
220120-625EA 61.28 USD
220120-625 220120-626
2,3-Dimethylaniline ≥99%
2,3-Dimethylaniline
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 222 °C (1013 hPa)
Melting Pt: -15 °C
Density: 0.99 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007732
CAS Number: 87-59-2
EINECS: 201-755-0
UN: 1711
ADR: 6.1,II
Merck Index: 12,10217

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR