3-(Dimethylamino)-1-propanol ≥99%

Synonyms: 3-(Dimethylamino)propan-1-ol, 3-Dimethylamino-1-propanol

D144401-100ML D144401-500ML
220120-883EA 45.02 USD
220120-883 220120-884
3-(Dimethylamino)-1-propanol ≥99%
3-(Dimethylamino)-1-propanol
Formula: C₅H₁₃NO
MW: 103.16 g/mol
Boiling Pt: 163 °C (1013 hPa)
Melting Pt: -35 °C
Density: 0.8878 g/cm³ (20 °C)
Flash Pt: 58 °C
MDL Number: MFCD00002945
CAS Number: 3179-63-3
EINECS: 221-659-2

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR