3-(Dimethylamino)phenol ≥97%

Synonyms: N,N-Dimethyl-3-hydroxyaniline, 3-Dimethylaminophenol

D144002-5G D144002-25G
220120-880EA 59.66 USD
220120-880 220120-879
3-(Dimethylamino)phenol ≥97%
3-(Dimethylamino)phenol
Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 265 °C (1013 hPa)
Melting Pt: 65…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00002264
CAS Number: 99-07-0
EINECS: 202-727-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR