2,6-Dimethoxytoluene ≥98%

Warning

D137200-25G D137200-100G
220121-049EA 210 USD
220121-049 220121-048
2,6-Dimethoxytoluene ≥98%
2,6-Dimethoxytoluene
Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Boiling Pt: 52 - 56 °C (0.5 -rr)
Melting Pt: 39 °C
MDL Number: MFCD00008374
CAS Number: 5673-07-4
EINECS: 227-131-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR