2,5-Dimethoxytetrahydrofuran cis- and trans-mixture

D137103-100G D137103-500G
220121-046EA 69.14 USD
220121-046 220121-047
2,5-Dimethoxytetrahydrofuran cis- and trans-mixture
2,5-Dimethoxytetrahydrofuran
Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Boiling Pt: 145 °C (1013 hPa)
Melting Pt: -45 °C
Density: 1.021 g/cm³ (20 °C)
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00005359
CAS Number: 696-59-3
EINECS: 211-797-1
UN: 3271
ADR: 3,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR