2,6-Dimethoxyphenol ≥99%

Warning

D135550-100G D135550-25G
220121-032EA 170.15 USD
220121-032 220121-033
2,6-Dimethoxyphenol ≥99%
2,6-Dimethoxyphenol
Formula: C₈H₁₀O₃
MW: 154.17 g/mol
Boiling Pt: 261 °C (1013 hPa)
Melting Pt: 52…56 °C
Storage Temperature: Ambient
MDL Number: MFCD00064434
CAS Number: 91-10-1
EINECS: 202-041-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR