2,6-Dimethoxynaphthalene

D134856-5G D134856-1G
220121-031EA 167.31 USD
220121-031 220121-030
2,6-Dimethoxynaphthalene
2,6-Dimethoxynaphthalene
Formula: C₁₂H₁₂O₂
MW: 188.23 g/mol
MDL Number: MFCD00004063
CAS Number: 5486-55-5
EINECS: 226-816-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR