3,4-Dimethoxybenzyl alcohol ≥96%

Synonyms: Veratryl alcohol, (3,4-Dimethoxyphenyl)methanol

D133000-25G D133000-100G
220120-949EA 73.23 USD
220120-949 220120-948
3,4-Dimethoxybenzyl alcohol ≥96%
3,4-Dimethoxybenzyl alcohol
Formula: C₉H₁₂O₃
MW: 168.19 g/mol
MDL Number: MFCD00004638
CAS Number: 93-03-8
EINECS: 202-212-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR