1,4-Diaminobutane ≥99%

Synonyms: Putrescine, 1,4-Butanediamine, Tetramethylendiamine

D13208-100G D13208-25G D13208-500G
220120-941EA 91.11 USD
220120-941 220120-942 220120-943
1,4-Diaminobutane ≥99%
1,4-Diaminobutane
Formula: C₄H₁₂N₂
MW: 88.15 g/mol
Boiling Pt: 158 °C (1013 hPa)
Melting Pt: 25…28 °C
Density: 0.88 g/cm³ (20 °C)
Flash Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD00008235
CAS Number: 110-60-1
EINECS: 203-782-3
UN: 2928
ADR: 6.1,II
Merck Index: 12,08134

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR