3,5-Dimethoxybenzoic acid ≥97%

D132004-100G D132004-25G
220120-127EA 598.4 USD
220120-127 220120-128
3,5-Dimethoxybenzoic acid ≥97%
3,5-Dimethoxybenzoic acid
Formula: C₉H₁₀O₄
MW: 182.18 g/mol
Boiling Pt: 341 °C (1013 hPa)
Melting Pt: 179 - 182 °C
Density: 1.423 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002502
CAS Number: 1132-21-4
EINECS: 214-473-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR