1,4-Dimethoxybenzene ≥99%

Synonyms: p-Dimethoxybenzene, Hydroquinone dimethyl ether

D131350-100G D131350-1KG D131350-500G
220120-114EA 29.51 USD
220120-114 220120-115 220120-116
1,4-Dimethoxybenzene ≥99%
1,4-Dimethoxybenzene
Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 212.6 °C (1013 hPa)
Melting Pt: 54…56 °C
Density: 0.874 g/cm³ (24.9 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008401
CAS Number: 150-78-7
EINECS: 205-771-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR