2,5-Dimethoxybenzaldehyde ≥99%

D130605-100G D130605-25G
220120-937EA 121.69 USD
220120-937 220120-938
2,5-Dimethoxybenzaldehyde ≥99%
2,5-Dimethoxybenzaldehyde
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 284 °C (1013 hPa)
Melting Pt: 49…51 °C
Storage Temperature: Ambient
MDL Number: MFCD00003314
CAS Number: 93-02-7
EINECS: 202-211-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR