2,4-Dimethoxybenzaldehyde ≥98%

D130400-100G D130400-25G
220120-934EA 236.92 USD
220120-934 220120-935
2,4-Dimethoxybenzaldehyde ≥98%
2,4-Dimethoxybenzaldehyde
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 308 °C (1013 hPa)
Melting Pt: 67…68 °C
Storage Temperature: Ambient
MDL Number: MFCD00003311
CAS Number: 613-45-6
EINECS: 210-342-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR