3,5-Dimethoxyaniline ≥98%

Warning

D130001-5G D130001-25G
220120-932EA 43.08 USD
220120-932 220120-931
3,5-Dimethoxyaniline ≥98%
3,5-Dimethoxyaniline
Formula: C₈H₁₁NO₂
MW: 153.18 g/mol
Boiling Pt: 303 °C (1013 hPa)
Melting Pt: 66…68 °C
Storage Temperature: Ambient
MDL Number: MFCD00008392
CAS Number: 10272-07-8
EINECS: 233-616-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR