Toluene-3,4-dithiol ≥90%, Technical Grade

Synonyms: 3,4-Toluenedithiol, 4-Methyl-1,2-dimercaptobenzene, 1,2-Dimercapto-4-methylbenzene, 4-Methylbenzene-1,2-dithiol

D129208-1ML D129208-5ML
220120-467EA 213.23 USD
220120-467 220120-468
Toluene-3,4-dithiol ≥90%, Technical Grade
Toluene-3,4-dithiol
Formula: C₇H₈S₂
MW: 156.27 g/mol
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 30…32 °C
Density: 1.18 g/cm³ (20 °C)
MDL Number: MFCD00004844
CAS Number: 496-74-2
EINECS: 207-828-3
Merck Index: 12,09669

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR