3,4-Diaminobenzoic acid ≥97%

Warning

D12600-5G D12600-25G D12600-100G
220120-448EA 36.51 USD
220120-448 220120-447 220120-446
3,4-Diaminobenzoic acid ≥97%
3,4-Diaminobenzoic acid
Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Boiling Pt: 354 °C (1013 hPa)
Melting Pt: 211 °C
Density: 1.544 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007726
CAS Number: 619-05-6
EINECS: 210-577-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR