3,3'-Diaminobenzidine ≥99%

Synonyms: [1,1'-Biphenyl]-3,3',4,4'-tetraamine, DAB, 3,3',4,4'-Biphenyltetramin, Biphenyl-3,3',4,4'-tetrayltetraamine

D12384-1G D12384-5G D12384-25G
220120-917EA 43.29 USD
220120-917 220120-919 220120-918
3,3'-Diaminobenzidine ≥99%
3,3'-Diaminobenzidine (DAB)
Formula: C₁₂H₁₄N₄
MW: 214.27 g/mol
MDL Number: MFCD00007725
CAS Number: 91-95-2
EINECS: 202-110-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR