1,4-Diiodobutane ≥99%

Warning

Synonyms: Tetramethylene diiodide

D122602-100G D122602-25G
220120-063EA 696.77 USD
220120-063 220120-064
1,4-Diiodobutane ≥99%
1,4-Diiodobutane
Formula: C₄H₈I₂
MW: 309.92 g/mol
Boiling Pt: 247 °C (1013 hPa)
Melting Pt: 5.8 °C
Density: 2.35 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001099
CAS Number: 628-21-7
EINECS: 211-032-1

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR