1,5-Dihydroxynaphthalene

Warning

Synonyms: 1,5-Naphthalenediol, Naphthalene-1,5-diol

D115606-500G D115606-100G
220120-271EA 210 USD
220120-271 220120-270
1,5-Dihydroxynaphthalene
1,5-Dihydroxynaphthalene
Formula: C₁₀H₈O₂
MW: 160.17 g/mol
Boiling Pt: 375 °C (1013 hPa)
Melting Pt: 180 °C
Density: 1.41 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003980
CAS Number: 83-56-7
EINECS: 201-487-4
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR