Diethylenetriaminepentaacetic acid ≥98%

Warning

Synonyms: N-Carboxymethyliminobis(ethylenenitrilo)tetra(acetic acid), Kestranal® Aza Pa, DTPA, Titriplex® V, Idranal V, Diethylenetriaminepentaacetic acid, 2,2',2'',2'''-((((Carboxymethyl)azanediyl)bis(ethane-2,1-diyl))bis(azanetriyl))tetraacetic acid, Diethylenetriamine-N,N,N',N'',N''-pentaacetic acid

D1133-1KG D1133-250G D1133-500G D1133-5G
220120-862EA 1002.46 USD
220120-862 220120-863 220120-864 220120-865
Diethylenetriaminepentaacetic acid ≥98%
Diethylenetriaminepentaacetic acid
Formula: C₁₄H₂₃N₃O₁₀
MW: 393.35 g/mol
Melting Pt: 220 - 222 °C
Density: 1.558 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004289
CAS Number: 67-43-6
EINECS: 200-652-8
UN: 3077
ADR: 9,III
Merck Index: 13,07202

Order Now

Specification Test Results

Assay 98% (titration)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR