4,4'-Dihydroxybenzophenone ≥99%

Synonyms: Bis(4-hydroxyphenyl) ketone

D110507-25G D110507-5G
220120-858EA 323 USD
220120-858 220120-859
4,4'-Dihydroxybenzophenone ≥99%
4,4'-Dihydroxybenzophenone
Formula: C₁₃H₁₀O₃
MW: 214.22 g/mol
Boiling Pt: 375 °C (1013 hPa)
Melting Pt: 214 - 217 °C
Density: 1.336 g/cm³ (20 °C)
MDL Number: MFCD00002358
CAS Number: 611-99-4
EINECS: 210-288-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR