3,5-Dihydroxybenzoic acid ≥97%

Synonyms: alpha-Resorcylic acid

D110000-1KG D110000-100G
220120-856EA 114.15 USD
220120-856 220120-855
3,5-Dihydroxybenzoic acid ≥97%
3,5-Dihydroxybenzoic acid
Formula: C₇H₆O₄
MW: 154.12 g/mol
Melting Pt: 236 - 238 °C
Flash Pt: ≥ 200 °C
Storage Temperature: Ambient
MDL Number: MFCD00002512
CAS Number: 99-10-5
EINECS: 202-730-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR