2,6-Dihydroxybenzoic acid ≥98%

D109606-100G D109606-25G
220120-853EA 58.15 USD
220120-853 220120-854
2,6-Dihydroxybenzoic acid ≥98%
2,6-Dihydroxybenzoic acid
Formula: C₇H₆O₄
MW: 154.12 g/mol
Boiling Pt: 339 °C (1013 hPa)
Melting Pt: 152 - 158 °C
Density: 1.58 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002462
CAS Number: 303-07-1
EINECS: 206-134-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR