3,4-Dihydroxybenzaldehyde ≥97%

Warning

Synonyms: Protocatechuic aldehyde, Protocatechualdehyde

D108405-100G D108405-25G D108405-5G
220120-846EA 157.23 USD
220120-846 220120-847 220120-848
3,4-Dihydroxybenzaldehyde ≥97%
3,4-Dihydroxybenzaldehyde
Formula: C₇H₆O₃
MW: 138.12 g/mol
Boiling Pt: 295 °C (1013 hPa)
Melting Pt: 150 - 155 °C
Density: 1.505 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003370
CAS Number: 139-85-5
EINECS: 205-377-7
Merck Index: 12,08079

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR