2,5-Dihydroxybenzaldehyde ≥98%

Warning

D108200-25G D108200-5G
220120-844EA 818.46 USD
220120-844 220120-845
2,5-Dihydroxybenzaldehyde ≥98%
2,5-Dihydroxybenzaldehyde
Formula: C₇H₆O₃
MW: 138.12 g/mol
Boiling Pt: 60…65 °C (12 mmHg)
Melting Pt: 99 °C
Density: 0.859 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003333
CAS Number: 1194-98-5
EINECS: 214-789-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR