2',4'-Dihydroxyacetophenone ≥99%

Warning

D107409-500G D107409-100G
220120-839EA 285.38 USD
220120-839 220120-838
2',4'-Dihydroxyacetophenone ≥99%
2',4'-Dihydroxyacetophenone
Formula: C₈H₈O₃
Boiling Pt: 336 °C (1013 hPa)
Melting Pt: 142 - 143 °C
Density: 1.18 g/cm³ (141 °C)
MDL Number: MFCD00002279
CAS Number: 89-84-9
EINECS: 201-945-3
Merck Index: 12,08308

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR