1,4-Difluorobenzene ≥99%

D102202-100G D102202-25G
220120-793EA 75.17 USD
220120-793 220120-794
1,4-Difluorobenzene ≥99%
1,4-Difluorobenzene
Formula: C₆H₄F₂
MW: 114.09 g/mol
Boiling Pt: 88 °C (1013 hPa)
Melting Pt: -13 °C
Density: 1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000344
CAS Number: 540-36-3
EINECS: 208-742-9
UN: 1993
ADR: 3,II
Merck Index: 13,03172

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR