2,4-Difluoroaniline ≥99%

Warning

D101400-10G D101400-100G D101400-25G
220120-789EA 56.43 USD
220120-789 220120-788 220120-790
2,4-Difluoroaniline ≥99%
2,4-Difluoroaniline
Formula: C₆H₅F₂N
MW: 129.11 g/mol
Boiling Pt: 170 °C (1013 hPa)
Melting Pt: –7 °C
Density: 1.268 g/cm³ (20 °C)
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00007648
CAS Number: 367-25-9
EINECS: 206-687-5
UN: 2941
ADR: 6.1,III
Merck Index: 13,03171

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR